Methyl (2R,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2R,4S)-4-Hydroxy-2-(methoxycarbonyl)pyrrolidine hydrochloride, tans-4-Hydroxy-D-proline methyl ester HCl, trans-4-Hydroxy-D-proline methyl ester hydrochloride, (2R,4S)-Methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride

A107793-250MG A107793-1G A107793-5G A107793-10G A107793-25G
103797-248EA 11 USD
103797-248 103823-016 77025-550 77309-868 77309-870 77570-458 77666-570
Methyl (2R,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%
Methyl (2R,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃·ClH
MW: 181.617 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00135747
CAS Number: 481704-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR