trans-4-Hydroxy-D-proline 97%

Supplier: Ambeed
Warning

Synonyms: (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, H-D-Hyp-OH, H-trans-D-Hyp-OH

A391394-1G A391394-5G A391394-10G A391394-250MG A391394-25G
103826-884EA 10 USD
103826-884 103837-922 76841-312 76841-314 77168-214 77168-216 77307-002
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline

Precaución: for Research Use Only

Formula: C₅H₉NO₃
MW: 131.131 g/mol
Boiling Pt: 355.2 °C (760 mm Hg)
Melting Pt: 261 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00065608
CAS Number: 3398-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR