Boc-D-trans-Hyp-OMe 97%

Supplier: Ambeed
Warning

A139174-10G A139174-1G A139174-25G A139174-5G A139174-100G A139174-250MG A139174-500G
103802-708EA 19 USD
103802-708 76719-288 77299-050 77299-052 77299-054 77600-016 77667-006 77667-008
Boc-D-trans-Hyp-OMe 97%
Boc-D-trans-Hyp-OMe

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
Boiling Pt: 335.2 °C (760 mm Hg)
Flash Pt: 156.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09954932
CAS Number: 135042-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR