(2R,4S)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A184346-100MG A184346-250MG A184346-1G
103815-746EA 42 USD
103815-746 103846-308 77012-074 77012-076 77301-902
(2R,4S)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride 95%
(2R,4S)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₁ClN₂O₄
MW: 280.75 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10568359
CAS Number: 1217446-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR