(R,R)-(-)-2,4-Pentanediol 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-(-)-2,4-Dihydroxy pentane, (2R,4R)-(-)-2,4-Pentanediol, (2R,4R)-Pentane-2,4-diol

A243740-1G A243740-250MG A243740-25G A243740-5G A243740-100G
103811-544EA 15 USD
103811-544 77090-574 77140-328 77140-330 77140-332 77685-684
(R,R)-(-)-2,4-Pentanediol 98%
(R,R)-(-)-2,4-Pentanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 174.021 °C (760 mm Hg)
Flash Pt: 72.784 °C
Storage Temperature: Ambient
MDL Number: MFCD00063893
CAS Number: 42075-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR