Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2R,4R)-Methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, N-D-cis-Hyp-OMe·HCl, cis-4-Hydroxy-D-proline methyl ester hydrochloride, (2R,4R)-4-Hydroxy-2-(methoxycarbonyl)pyrrolidine hydrochloride

A223355-10G A223355-1G A223355-5G A223355-25G A223355-100G
103819-028EA 15 USD
103819-028 103849-910 77030-880 77088-528 77088-530 77303-648 77667-854
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃·ClH
MW: 181.617 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082536
CAS Number: 114676-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR