2'-Deoxyadenosine 98%

Supplier: Ambeed
Danger

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

A159248-10G A159248-1G A159248-25G A159248-5G
103843-452EA 22 USD
103843-452 76689-748 76689-750 77319-326 77319-328
2'-Deoxyadenosine 98%
2'-Deoxyadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₃
MW: 251.24 g/mol
Melting Pt: 187…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056003
CAS Number: 958-09-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR