(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-α-D-arabinofuranose, 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-α-D-arabinofuranose

A134316-100G A134316-10G A134316-1G A134316-25G A134316-5G A134316-250MG A134316-500G
103802-000EA 11 USD
103802-000 76996-570 76996-572 77314-730 77314-732 77314-734 77670-926 77670-928
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate

Precaución: for Research Use Only

Formula: C₂₆H₂₁FO₇
MW: 464.44 g/mol
Boiling Pt: 584.1 °C (760 mm Hg)
Melting Pt: 74…77 °C
Flash Pt: 295.665 °C
Storage Temperature: Ambient
MDL Number: MFCD00083339
CAS Number: 97614-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR