Tri-O-acetyl-D-glucal 98%

Supplier: Ambeed
Warning

A136632-100G A136632-10G A136632-1G A136632-500G A136632-25G A136632-5G
76767-554EA 99.73 USD
76767-554 76767-556 76767-558 76767-560 77315-244 77315-246
Tri-O-acetyl-D-glucal 98%
Tri-O-acetyl-D-glucal

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₇
MW: 272.25 g/mol
Boiling Pt: 343.1 °C (760 mm Hg)
Melting Pt: 53…55 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00063253
CAS Number: 2873-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR