N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid, N-Fmoc-D-threonine, Fmoc-D-Thr-OH

A242040-1G A242040-5G
103811-450EA 8 USD
103811-450 103851-260 77090-422 77090-424
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
Boiling Pt: 596.5 °C (760 mm Hg)
Melting Pt: 88…92 °C
Flash Pt: 314.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153365
CAS Number: 157355-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR