Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside 97%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1-thio-β-D-glucopyranoside tetraacetate, (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate, Phenyl 2,3,4,6-tetra-o-acetyl-1-thio-β-D-glucopyranoside

A100973-100MG A100973-250MG A100973-25G A100973-1G A100973-5G
103795-728EA 21 USD
103795-728 103821-420 76717-512 76717-514 76717-516 77282-624 77282-626
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside 97%
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₂₀H₂₄O₉S
MW: 440.46 g/mol
Boiling Pt: 514.6 °C (760 mm Hg)
Melting Pt: 116…119 °C
Flash Pt: 247.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00135159
CAS Number: 23661-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR