(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloropurine riboside
A569930-25G
A569930-5G
A569930-100G
103828-798EA
6
USD
103828-798
103839-942
77307-846
77307-848
77749-173
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 98%
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
Precaución: for Research Use Only
Formula:
C₁₀H₁₁ClN₄O₄ MW: 286.67 g/mol Boiling Pt: 614.8 °C (760 mm Hg) Melting Pt: 158…162 °C Flash Pt: 325.6 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005738 CAS Number: 5399-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...