8-Bromoadenosine 98%

Supplier: Ambeed
Warning

Synonyms: 8-Bromoadenine-9-β-D-ribofuranoside, 6-Amino-8-bromopurine riboside, (2R,3R,4S,5R)-2-(6-Amino-8-bromo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

A130841-250MG A130841-1G
103801-484EA 10 USD
103801-484 76681-624 77313-954
8-Bromoadenosine 98%
8-Bromoadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN₅O₄
MW: 346.15 g/mol
Boiling Pt: 717.7 °C (760 mm Hg)
Melting Pt: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00005733
CAS Number: 2946-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR