(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 97%

Supplier: Ambeed
Warning

A100186-250MG A100186-1G A100186-100MG
103795-286EA 59 USD
103795-286 103821-000 77270-140 77296-590 77747-747
(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 97%
(2R,3R,4S,5R)-2-(2,6-Dichloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₄O₄
MW: 321.12 g/mol
Boiling Pt: 589.3 °C (760 mm Hg)
Melting Pt: >190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411342
CAS Number: 13276-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR