D-(+)-Galactosamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

A128462-100G A128462-10G A128462-500G A128462-1G A128462-25G A128462-5G
103801-128EA 5 USD
103801-128 76996-210 76996-212 76996-214 77313-438 77313-440 77313-442
D-(+)-Galactosamine hydrochloride 98%
D(+)-Galactosamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
Boiling Pt: 449 °C (760 mm Hg)
Flash Pt: 225.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00135830
CAS Number: 1772-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR