Tri-O-acetyl-D-galactal 95%
Supplier: Ambeed
|
Warning
|
A113736-100G
A113736-10G
A113736-250MG
A113736-25G
A113736-1G
A113736-5G
103798-282EA
10
USD
103798-282
103824-126
77271-064
77271-066
77271-068
77271-070
77310-938
77310-940
Tri-O-acetyl-D-galactal 95%
Tri-O-acetyl-D-galactal
Precaución: for Research Use Only
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol Boiling Pt: 343.1 °C (760 mm Hg) Melting Pt: 34…38 °C Flash Pt: 149.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064092 CAS Number: 4098-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...