(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R)-3-((R)-1-((tert-Butyldimethylsilyl)oxy)ethyl)-4-oxoazetidin-2-yl acetate, (3R,4R)-(+)-4-Acetoxy-3-[1-(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone
A147479-100G
A147479-10G
A147479-25G
A147479-5G
103841-888EA
13
USD
103841-888
76719-588
76719-590
77299-582
77299-584
(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone 97%
(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone
Precaución: for Research Use Only
Formula:
C₁₃H₂₅NO₄Si MW: 287.43 g/mol Melting Pt: 107…109 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077636 CAS Number: 76855-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...