(-)-Di-p-anisoyl-L-tartaric acid 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid, (-)-Bis(4-methoxybenzoyl)-L-tartaric Acid

A295764-25G A295764-100G A295764-10G A295764-500G A295764-5G
103835-204EA 7 USD
103835-204 76837-868 77289-878 77289-880 77289-882 77305-678
(-)-Di-p-anisoyl-L-tartaric acid 97%
(-)-Di-p-anisoyl-L-tartaric acid

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₁₀
Boiling Pt: 681.554 °C (760 mm Hg)
Melting Pt: 193…195 °C
Flash Pt: 241.638 °C
Storage Temperature: Ambient
MDL Number: MFCD02682986
CAS Number: 50583-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR