(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol with methanesulfonate 97%
Supplier: Ambeed
|
Warning
|
A125292-25G
A125292-5G
103800-664EA
15
USD
103800-664
77246-304
77312-818
(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol with methanesulfonate 97%
(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol with methanesulfonate
Precaución: for Research Use Only
Formula:
C₁₃H₁₇F₂N₃O₅S Storage Temperature: Ambient |
MDL Number:
MFCD28964178 CAS Number: 1175536-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...