(H-Cys-OtBu)₂·2 HCl 95%

Supplier: Ambeed
Warning

Synonyms: (2R,2'R)-Di-tert-butyl 3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride

A161063-100G A161063-10G A161063-1G A161063-5G
103805-448EA 8 USD
103805-448 103843-686 77138-162 77138-164 77300-510 77300-512
(H-Cys-OtBu)₂·2 HCl 95%
(H-Cys-OtBu)₂·2 HCl

Precaución: for Research Use Only

Formula: C₁₄H₃₀Cl₂N₂O₄S₂
MW: 425.424 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237452
CAS Number: 38261-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR