(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid) 96%

Supplier: Ambeed
Warning

Synonyms: (Fmoc-Cys-OH)₂

A884411-5G A884411-100G A884411-10G A884411-25G A884411-1G
103841-320EA 33 USD
103841-320 76845-588 77147-020 77147-022 77147-024 77308-486
(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid) 96%
(2R,2'R)-3,3'-Disulfanediylbis(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid)

Precaución: for Research Use Only

Formula: C₃₆H₃₂N₂O₈S₂
MW: 684.778 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237447
CAS Number: 135273-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR