(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine 97%

Supplier: Ambeed
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene

A212428-100MG A212428-250MG A212428-5G
103818-198EA 65 USD
103818-198 77237-418 77237-420 77237-422
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine 97%
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine

Precaución: for Research Use Only

Formula: C₄₀H₄₀FeP₂
MW: 638.5400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426997
CAS Number: 184095-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR