(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid 96%

Supplier: Ambeed
Warning

A214510-100MG A214510-250MG A214510-1G
103818-410EA 207 USD
103818-410 77030-606 77030-608 77303-356
(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid 96%
(2-Oxo-2,3-dihydrobenzo[d]oxazol-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₄
MW: 178.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072745
CAS Number: 1016644-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR