β-Nitrostyrene 95%

Supplier: Ambeed
Warning

Synonyms: (1-Nitro-2-phenyl)ethylene, (2-Nitrovinyl)benzene, Nitrostyrene, ω-Nitrostyrene

A114800-100G A114800-10G A114800-25G A114800-1G A114800-5G
103798-480EA 6 USD
103798-480 103824-322 76781-326 76781-328 76781-330 77297-328 77297-330
β-Nitrostyrene 95%
β-Nitrostyrene

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.14 g/mol
Boiling Pt: 250…260 °C at N/A mmHg
Melting Pt: 55…58 °C
Flash Pt: 117.5 °C
Storage Temperature: Freezer
CAS Number: 5153-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR