(2-Nitrophenyl)hydrazine hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Nitrophenylhydrazine hydrochloride

A153750-25G A153750-5G
103842-736EA 7 USD
103842-736 77300-028 77318-416
(2-Nitrophenyl)hydrazine hydrochloride 98%
(2-Nitrophenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈ClN₃O₂
Boiling Pt: 314.3 °C (760 mm Hg)
Flash Pt: 143.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060205
CAS Number: 6293-87-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR