2-Methyl-4-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Methylpyridin-4-yl)boronic acid, 2-Methylpyridine-4-boronic acid

A405189-1G A405189-10G A405189-250MG
103827-116EA 18 USD
103827-116 103838-158 76841-646 77168-326 77168-328
2-Methyl-4-pyridineboronic acid 97%
2-Methyl-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂
MW: 136.9442 g/mol
Boiling Pt: 311.2 °C (760 mm Hg)
Flash Pt: 142 °C
Storage Temperature: Freezer
MDL Number: MFCD03095364
CAS Number: 579476-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR