2-Methyl-3-pyridineboronic acid hydrochloride 97%

Supplier: Ambeed
Warning

A136965-250MG A136965-1G A136965-5G
103802-386EA 31 USD
103802-386 76823-264 77315-318 77315-320
2-Methyl-3-pyridineboronic acid hydrochloride 97%
2-Methyl-3-pyridineboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06659489
CAS Number: 1072952-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR