2-Methyl-1-phenylpropene 97%

Supplier: Ambeed
Danger

Synonyms: (2-Methylprop-1-en-1-yl)benzene, (2-Methylpropenyl)benzene

A441670-1G A441670-25G A441670-5G A441670-100MG A441670-10G A441670-250MG
103827-676EA 17 USD
103827-676 103838-746 76842-510 76842-512 76842-514 77264-996 77264-998 77265-000
2-Methyl-1-phenylpropene 97%
2-Methyl-1-phenylpropene

Precaución: for Research Use Only

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 187…188 °C (760 mm Hg)
Melting Pt: 5048 °C
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008899
CAS Number: 768-49-0
UN: 3295
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR