(2-Methoxy-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

A198472-10G A198472-250MG A198472-1G A198472-25G
103817-020EA 13 USD
103817-020 103847-646 76999-234 76999-236 77085-672 77085-674
(2-Methoxy-3-pyridinyl)methanol 97%
(2-Methoxy-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Boiling Pt: 241.8 °C (760 mm Hg)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD09880283
CAS Number: 112197-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR