(2-Methoxypyridin-3-yl)methanamine 97%

Supplier: Ambeed
Warning

A281544-1G A281544-250MG A281544-5G
103813-558EA 58 USD
103813-558 103853-508 76837-204 76977-420 77305-400
(2-Methoxypyridin-3-yl)methanamine 97%
(2-Methoxypyridin-3-yl)methanamine

Precaución: For research use only

Formula: C₇H₁₀N₂O
Boiling Pt: 224.7±25.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09740382
CAS Number: 354824-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR