2-Methoxybenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol

A123268-100G A123268-25G A123268-500G A123268-5G
77258-146EA 128.75 USD
77258-146 77258-148 77258-150 77312-498
2-Methoxybenzyl alcohol 97%
2-Methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 249 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004611
CAS Number: 612-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR