(2-Methoxyethyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

A252348-100MG A252348-10G A252348-250MG A252348-1G A252348-25G A252348-5G A252348-100G
103812-038EA 8 USD
103812-038 103851-882 76826-990 76826-992 76826-994 77091-294 77091-296 77091-298 77683-756
(2-Methoxyethyl)triphenylphosphonium bromide 97%
(2-Methoxyethyl)triphenylphosphonium bromide

Precaución: For research use only.

Formula: C₂₁H₂₂BrOP
MW: 401.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00195808
CAS Number: 55894-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR