(2-Methoxy-6-methylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-6-methylphenylboronic acid

A175974-100MG A175974-1G A175974-250MG
103807-104EA 191 USD
103807-104 103845-416 77096-626 77301-410 77685-395
(2-Methoxy-6-methylphenyl)boronic acid 95%
(2-Methoxy-6-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11043444
CAS Number: 1567218-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR