2-Methoxy-4,6-dimethylbenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Methoxy-4,6-dimethylphenyl)boronic acid

A100927-10G A100927-250MG A100927-1G A100927-5G
103795-706EA 95 USD
103795-706 103821-394 77245-086 77245-088 77282-606 77282-608
2-Methoxy-4,6-dimethylbenzeneboronic acid 97%
2-Methoxy-4,6-dimethylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04037222
CAS Number: 355836-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR