2-Isopropylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropylbenzeneboronic acid, (2-Isopropylphenyl)boronic acid

A321103-1G A321103-25G A321103-5G A321103-100G A321103-10G
103825-264EA 9 USD
103825-264 103836-196 76839-086 76839-088 76839-090 77251-142 77251-144
2-Isopropylphenylboronic acid 98%
2-Isopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Melting Pt: 85…86 °C
Flash Pt: 133.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411937
CAS Number: 89787-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR