(2-Isopropoxypyrimidin-5-yl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropoxy-5-pyrimidineboronic acid

A178456-100MG A178456-1G A178456-5G A178456-250MG
103807-336EA 58 USD
103807-336 103845-674 76985-668 77301-556 77301-558 77683-429
(2-Isopropoxypyrimidin-5-yl)boronic acid 97%
(2-Isopropoxypyrimidin-5-yl)boronic acid

Precaución: For research use only.

Formula: C₇H₁₁BN₂O₃
MW: 181.98 g/mol
Boiling Pt: 364.3±52.0 °C (760 mm Hg)
Flash Pt: 174.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16618883
CAS Number: 870521-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR