2-Isopropoxy-3-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropoxypyridine-3-boronic acid, (2-Isopropoxypyridin-3-yl)boronic acid

A585278-100MG A585278-5G
103828-882EA 22 USD
103828-882 103840-028 76819-266 77307-876
2-Isopropoxy-3-pyridineboronic acid 95%
2-Isopropoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181.0000 g/mol
Boiling Pt: 336 °C (760 mm Hg)
Flash Pt: 157 °C
Storage Temperature: Freezer
MDL Number: MFCD11855840
CAS Number: 1150114-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR