(2-Iodophenyl)methanamine 95%

Supplier: Ambeed
Danger

A252932-1G A252932-100MG A252932-250MG
103812-064EA 74 USD
103812-064 103851-912 77091-336 77262-028 77262-030
(2-Iodophenyl)methanamine 95%
(2-Iodophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₈IN
Boiling Pt: 270 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00047929
CAS Number: 39959-51-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR