2-Formyl-3-thiopheneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid

A101967-10G A101967-250MG A101967-5G
103796-232EA 22 USD
103796-232 103821-930 77008-354 77008-356 77308-812
2-Formyl-3-thiopheneboronic acid 97%
2-Formyl-3-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Boiling Pt: 396.7 °C (760 mm Hg)
Melting Pt: 167…193 °C
Storage Temperature: Freezer
MDL Number: MFCD01075678
CAS Number: 4347-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR