2-Formylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane

A304580-100G A304580-10G A304580-1G A304580-500G A304580-25G A304580-5G
103835-588EA 8 USD
103835-588 76772-684 76772-686 76772-688 76772-690 76838-282 76838-284
2-Formylphenylboronic acid 98%
2-Formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₃
Boiling Pt: 355.7 °C (760 mm Hg)
Melting Pt: 115…120 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00151822
CAS Number: 40138-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR