2-formyl-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A118444-100MG A118444-10G A118444-1G A118444-5G A118444-250MG
103799-060EA 62 USD
103799-060 76983-092 76983-094 77297-600 77297-602 77610-972
2-formyl-3-methoxyphenylboronic acid 98%
2-formyl-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
Boiling Pt: 408.3±55.0 °C (760 mm Hg)
Flash Pt: 200.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319042
CAS Number: 958030-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR