2-Fluoro-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid

A416748-1G A416748-25G A416748-5G A416748-100G A416748-10G A416748-250MG
103827-328EA 11 USD
103827-328 103838-374 76841-954 76841-956 76841-958 77016-408 77016-410 77016-412
2-Fluoro-4-pyridineboronic acid 98%
2-Fluoro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Boiling Pt: 349.3 °C (760 mm Hg)
Melting Pt: 160 °C
Flash Pt: 151.5 °C
Storage Temperature: Freezer
MDL Number: MFCD04112534
CAS Number: 401815-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR