2-Fluoro-6-hydroxyphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-6-hydroxyphenyl)boronic acid

A112058-100G A112058-100MG A112058-10G A112058-250MG A112058-25G A112058-1G A112058-5G
103797-944EA 9 USD
103797-944 103823-776 77008-954 77008-956 77008-958 77008-960 77008-962 77310-580 77310-582
2-Fluoro-6-hydroxyphenylboronic acid 95%
2-Fluoro-6-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.919 g/mol
Boiling Pt: 333.3±52.0 °C (760 mm Hg)
Flash Pt: 155.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17214242
CAS Number: 1256345-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR