2-Fluoro-5-hydroxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-hydroxyphenyl)boronic acid

A143572-10G A143572-250MG A143572-25G A143572-5G
103803-298EA 37 USD
103803-298 76871-334 76871-336 77299-350 77299-352
2-Fluoro-5-hydroxyphenylboronic acid 97%
2-Fluoro-5-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.919 g/mol
Boiling Pt: 386.5±52.0 °C (760 mm Hg)
Flash Pt: 187.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11855845
CAS Number: 1150114-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR