2-Fluoro-5-(trifluoromethoxy)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Fluoro-5-(trifluoromethoxy)phenyl)boronic acid
A349018-1G
A349018-10G
A349018-250MG
103826-008EA
55
USD
103826-008
103836-998
76840-082
77278-126
77278-128
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid 98%
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BF₄O₃ Boiling Pt: 279.9 °C (760 mm Hg) Flash Pt: 123.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09027234 CAS Number: 881402-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...