2-Fluoro-3-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

A193379-25G A193379-5G A193379-100G A193379-10G A193379-1G
103816-526EA 8 USD
103816-526 103847-140 77085-042 77085-044 77192-038 77192-040 77302-338
2-Fluoro-3-methoxyphenylboronic acid 97%
2-Fluoro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
Boiling Pt: 326.8 °C (760 mm Hg)
Melting Pt: 117…122 °C
Flash Pt: 151.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02094728
CAS Number: 352303-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR