2-Fluoro-3-hydroxybenzeneboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Fluoro-3-hydroxyphenyl)boronic acid, 2-Fluoro-3-hydroxyphenylboronic acid, 3-Borono-2-fluorophenol
A102687-100MG
A102687-10G
A102687-250MG
A102687-5G
103796-410EA
12
USD
103796-410
103822-108
76869-200
76869-202
76869-204
77308-978
2-Fluoro-3-hydroxybenzeneboronic acid 97%
2-Fluoro-3-hydroxybenzeneboronic acid
Precaución: for Research Use Only
Formula:
C₆H₆BFO₃ MW: 155.919 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06797222 CAS Number: 855230-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...