(2-Ethynylphenyl)methanamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A460955-250MG A460955-100MG
103827-922EA 152 USD
103827-922 103839-000 77613-426 77684-251
(2-Ethynylphenyl)methanamine hydrochloride ≥95%
(2-Ethynylphenyl)methanamine hydrochloride

Precaución: For research use only.

Formula: C₉H₁₀ClN
MW: 167.63 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26793460
CAS Number: 2389017-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR