2-Ethoxy-5-pyrimidineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Ethoxypyrimidin-5-yl)boronic acid

A237977-10G A237977-5G
103811-190EA 40 USD
103811-190 103850-974 76826-664 77304-096
2-Ethoxy-5-pyrimidineboronic acid 95%
2-Ethoxy-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₆H₉BN₂O₃
MW: 167.9600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375134
CAS Number: 1003043-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR