(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: (Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide

A118642-25G A118642-500G A118642-1000G A118642-5G A118642-100G
103808-188EA 235 USD
103808-188 77136-386 77136-388 77136-390 77297-616 77297-618 77566-172
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide

Precaución: For research use only.

Formula: C₂₂H₂₂BrO₂P
MW: 429.30 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011835
CAS Number: 1530-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR